2,3-Dicyanopyrazine 99%

Supplier: Ambeed
Danger

Synonyms: Pyrazine-2,3-dicarbonitrile

A447325-1G A447325-25G A447325-5G
77252-728EA 12.36 USD
77252-728 77252-730 77252-732
2,3-Dicyanopyrazine 99%
2,3-Dicyanopyrazine

Precaución: for Research Use Only

Formula: C₆H₂N₄
MW: 130.1069 g/mol
MDL Number: MFCD00059771
CAS Number: 13481-25-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR