1-Boc-Azepane-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A447138-250MG A447138-1G A447138-5G
77241-020EA 86 USD
77241-020 77551-346 77746-916
1-Boc-Azepane-4-carboxylic acid 97%
1-Boc-Azepane-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD10687156
CAS Number: 868284-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR