(Z)-2-(2-Aminothiazol-4-yl)-2-((2-methoxy-2-oxoethoxy)imino)acetic acid 95%

Supplier: Ambeed
Danger

A447033-100G A447033-1G A447033-25G A447033-500G A447033-5G
77252-718EA 96 USD
77252-718 77252-720 77252-722 77252-724 77252-726
(Z)-2-(2-Aminothiazol-4-yl)-2-((2-methoxy-2-oxoethoxy)imino)acetic acid 95%
(Z)-2-(2-Aminothiazol-4-yl)-2-((2-methoxy-2-oxoethoxy)imino)acetic acid

Precaución: for Research Use Only

Formula: C₈H₉N₃O₅S
MDL Number: MFCD07782100
CAS Number: 80544-17-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR