(5aS,10bR)-2-(2,4,6-Trichlorophenyl)-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A447011-100MG A447011-1G A447011-250MG A447011-50MG
77252-710EA 87.55 USD
77252-710 77252-712 77252-714 77252-716
(5aS,10bR)-2-(2,4,6-Trichlorophenyl)-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate 97%
(5aS,10bR)-2-(2,4,6-Trichlorophenyl)-4,5a,6,10b-tetrahydro-2H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-11-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₈H₁₃BCl₃F₄N₃O
MW: 480.47 g/mol
CAS Number: 1214711-48-4
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR