(R)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-diphenyl-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A446836-100MG
A446836-1G
A446836-250MG
A446836-5G
77252-698EA
78.28
USD
77252-698
77252-700
77252-702
77252-704
(R)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-diphenyl-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(R)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-diphenyl-[1,1'-binaphthalene]-2,2'-diol
Precaución: for Research Use Only
Formula:
C₃₂H₃₀O₂ MW: 446.60 g/mol |
CAS Number:
396134-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...