(E)-5-Chloro-3-((5-(3-(4-methyl-1,4-diazepane-1-carbonyl)phenyl)furan-2-yl)methylene)indolin-2-one hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: CX-6258 HCl

A446749-1MG A446749-5MG
77265-052EA 19.57 USD
77265-052 77265-054
(E)-5-Chloro-3-((5-(3-(4-methyl-1,4-diazepane-1-carbonyl)phenyl)furan-2-yl)methylene)indolin-2-one hydrochloride 98%
(E)-5-Chloro-3-((5-(3-(4-methyl-1,4-diazepane-1-carbonyl)phenyl)furan-2-yl)methylene)indolin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₂₆H₂₅Cl₂N₃O₃
MW: 498.40 g/mol
MDL Number: MFCD28137787
CAS Number: 1353859-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR