Serabelisib 98+%

Supplier: Ambeed
Warning

Synonyms: TAK-117), [6-(2-amino-5-benzoxazolyl)imidazo[1,2-a]pyridin-3-yl]-4-morpholinyl-methanone, (6-(2-Aminobenzo[d]oxazol-5-yl)imidazo[1,2-a]pyridin-3-yl)(morpholino)methanone, MLN1117, INK1117

A446445-25MG A446445-5MG A446445-10MG A446445-1MG
77241-006EA 25.75 USD
77241-006 77241-008 77616-912 77616-914
Serabelisib 98+%
Serabelisib

Precaución: for Research Use Only

Formula: C₁₉H₁₇N₅O₃
MW: 363.34 g/mol
MDL Number: MFCD15146372
CAS Number: 1268454-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR