7,4'-Di-O-methylapigenin 95%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-7-methoxy-2-(4-methoxyphenyl)-4H-chromen-4-one

A446385-1MG A446385-2MG A446385-5MG
77291-648EA 62.83 USD
77291-648 77291-650 77291-652
7,4'-Di-O-methylapigenin 95%
7,4'-Di-O-methylapigenin

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₅
MW: 298.29 g/mol
MDL Number: MFCD00017446
CAS Number: 5128-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR