2,6-Difluoronicotinaldehyde 98%

Supplier: Ambeed
Warning

A445948-5G A445948-25G
77252-690EA 30 USD
77252-690 77746-913
2,6-Difluoronicotinaldehyde 98%
2,6-Difluoronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃F₂NO
MDL Number: MFCD08277633
CAS Number: 155601-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR