2-Chloro-5-(trifluoromethyl)phenyl isocyanate 97%

Supplier: Ambeed
Danger

A445882-1G A445882-5G
77265-048EA 17 USD
77265-048 77265-050
2-Chloro-5-(trifluoromethyl)phenyl isocyanate 97%
2-Chloro-5-(trifluoromethyl)phenyl isocyanate

Precaución: for Research Use Only

Formula: C₈H₃ClF₃NO
MDL Number: MFCD00037029
CAS Number: 50528-86-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR