3-Chloro-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one 97%

Supplier: Ambeed
Warning

A445680-10G A445680-1G A445680-250MG A445680-25G A445680-5G
77252-680EA 396.55 USD
77252-680 77252-682 77252-684 77252-686 77252-688
3-Chloro-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one 97%
3-Chloro-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one

Precaución: for Research Use Only

Formula: C₁₇H₁₃ClO₂
MW: 284.74 g/mol
MDL Number: MFCD29917576
CAS Number: 1378388-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR