1-Ethyl-5-iodo-2-methyl-4-nitro-1H-imidazole 97%

Supplier: Ambeed
Warning

A445408-100MG A445408-1G A445408-250MG
77241-000EA 74.16 USD
77241-000 77241-002 77241-004
1-Ethyl-5-iodo-2-methyl-4-nitro-1H-imidazole 97%
1-Ethyl-5-iodo-2-methyl-4-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₆H₈IN₃O₂
MW: 281.05 g/mol
MDL Number: MFCD00100439
CAS Number: 35681-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR