2-Iminothiolane hydrochloride (Traut's Reagent) 95%

Supplier: Ambeed
Warning

A445403-100MG A445403-1G A445403-250MG A445403-5G
77265-040EA 22 USD
77265-040 77265-042 77265-044 77265-046
2-Iminothiolane hydrochloride (Traut's Reagent) 95%
2-Iminothiolane hydrochloride (Traut's Reagent)

Precaución: for Research Use Only

Formula: C₄H₈ClNS
MDL Number: MFCD00039013
CAS Number: 4781-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR