(4R,4'R)-2,2'-(Cyclobutane-1,1-diyl)bis(4-phenyl-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A445203-100MG A445203-1G A445203-250MG
77265-032EA 111.24 USD
77265-032 77265-034 77265-036
(4R,4'R)-2,2'-(Cyclobutane-1,1-diyl)bis(4-phenyl-4,5-dihydrooxazole) 98%
(4R,4'R)-2,2'-(Cyclobutane-1,1-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₂H₂₂N₂O₂
MW: 346.42 g/mol
CAS Number: 1246401-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR