3-Fluoro-4-propoxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

A445027-1G A445027-250MG A445027-25G A445027-5G
77279-158EA 15 USD
77279-158 77279-160 77279-162 77279-164
3-Fluoro-4-propoxybenzeneboronic acid 98%
3-Fluoro-4-propoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
MDL Number: MFCD06411285
CAS Number: 192376-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR