2,6-Bis(2-pyridyl)-4(1H)-pyridone 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Di(2-pyridyl)-4(1H)-pyridone, [2,2':6',2''-Terpyridin]-4'(1'H)-one

A444427-1G A444427-250MG A444427-5G A444427-100MG
77252-660EA 105.06 USD
77252-660 77252-662 77252-664 77616-908
2,6-Bis(2-pyridyl)-4(1H)-pyridone 98%
2,6-Bis(2-pyridyl)-4(1H)-pyridone

Precaución: for Research Use Only

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
MDL Number: MFCD01321386
CAS Number: 128143-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR