1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

A444144-100G A444144-10G A444144-1G A444144-25G A444144-5G
77240-980EA 129 USD
77240-980 77240-982 77240-984 77240-986 77746-908
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate 98%
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₇H₁₁F₃N₂O₃S
MDL Number: MFCD00210008
CAS Number: 145022-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR