racemic-4-Bromo[2.2]paracyclophane 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo[2.2]paracyclophane

A443797-100MG A443797-1G A443797-250MG A443797-5G
77291-626EA 19.57 USD
77291-626 77291-628 77291-630 77291-632
racemic-4-Bromo[2.2]paracyclophane 98%
racemic-4-Bromo[2.2]paracyclophane

Precaución: for Research Use Only

Formula: C₁₆H₁₅Br
MW: 287.1943 g/mol
MDL Number: MFCD00187358
CAS Number: 1908-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR