Chlorodiisopropyl(phenethyl)silane 95%

Supplier: Ambeed
Danger

A443510-100MG A443510-250MG A443510-5G
77252-654EA 19.57 USD
77252-654 77252-656 77252-658
Chlorodiisopropyl(phenethyl)silane 95%
Chlorodiisopropyl(phenethyl)silane

Precaución: for Research Use Only

Formula: C₁₄H₂₃ClSi
MW: 254.87 g/mol
MDL Number: MFCD01320966
CAS Number: 151613-24-0
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR