4-(Diphenylamino)benzonitrile 97%

Supplier: Ambeed
Warning

A443457-1G A443457-250MG
77291-622EA 65 USD
77291-622 77291-624
4-(Diphenylamino)benzonitrile 97%
4-(Diphenylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₁₉H₁₄N₂
MDL Number: MFCD09833579
CAS Number: 20441-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR