3-Bromo-6,7-dihydro-5H-cyclopenta[b]pyridine 95%

Supplier: Ambeed
Warning

A443296-100MG A443296-1G A443296-250MG
77279-146EA 64.89 USD
77279-146 77279-148 77279-150
3-Bromo-6,7-dihydro-5H-cyclopenta[b]pyridine 95%
3-Bromo-6,7-dihydro-5H-cyclopenta[b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈BrN
MDL Number: MFCD08706061
CAS Number: 158331-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR