Pf-06282999 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(6-(5-Chloro-2-methoxyphenyl)-4-oxo-2-thioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamide

A443096-10MG A443096-5MG A443096-1MG A443096-50MG
77265-010EA 47 USD
77265-010 77265-012 77616-900 77748-903
Pf-06282999 98+%
Pf-06282999

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN₃O₃S
MW: 325.77 g/mol
MDL Number: MFCD30533689
CAS Number: 1435467-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR