2,2,5-Trimethyl-1,3-dioxane-4,6-dione 97%

Supplier: Ambeed
Warning

A442437-1G A442437-5G A442437-100G A442437-10G A442437-25G
77252-638EA 5 USD
77252-638 77252-640 77594-382 77746-903 77746-904
2,2,5-Trimethyl-1,3-dioxane-4,6-dione 97%
2,2,5-Trimethyl-1,3-dioxane-4,6-dione

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MDL Number: MFCD00006637
CAS Number: 3709-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR