4,4',4''-Tri-tert-butyl-2,2':6',2''-terpyridine 97%

Supplier: Ambeed
Warning

A442218-100MG A442218-1G A442218-250MG A442218-5G
77291-614EA 30 USD
77291-614 77291-616 77291-618 77748-902
4,4',4''-Tri-tert-butyl-2,2':6',2''-terpyridine 97%
4,4',4''-Tri-tert-butyl-2,2':6',2''-terpyridine

Precaución: for Research Use Only

Formula: C₂₇H₃₅N₃
MDL Number: MFCD02093731
CAS Number: 115091-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR