5-Chloro-4-fluoropyridin-2-amine 97%

Supplier: Ambeed
Warning

A442195-250MG A442195-5G
77265-006EA 47 USD
77265-006 77568-878
5-Chloro-4-fluoropyridin-2-amine 97%
5-Chloro-4-fluoropyridin-2-amine

Precaución: For research use only.

Formula: C₅H₄ClFN₂
MDL Number: MFCD22547000
CAS Number: 1393574-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR