4-Bromo-2,6-difluorophenylboronic acid 95%

Supplier: Ambeed
Warning

A442167-1G A442167-25G A442167-5G
77291-608EA 9 USD
77291-608 77291-610 77291-612
4-Bromo-2,6-difluorophenylboronic acid 95%
4-Bromo-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrF₂O₂
MDL Number: MFCD03095369
CAS Number: 352535-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR