2,6-Dichloro-3-nitropyridine 98%

Supplier: Ambeed
Warning

A441790-100G A441790-10G A441790-25G A441790-500G A441790-5G
77279-126EA 26 USD
77279-126 77279-128 77279-130 77279-132 77279-134
2,6-Dichloro-3-nitropyridine 98%
2,6-Dichloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂N₂O₂
MDL Number: MFCD00006234
CAS Number: 16013-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR