2-Iodo-5-methoxyaniline 97%

Supplier: Ambeed
Warning

A441716-1G A441716-250MG
77265-002EA 17 USD
77265-002 77265-004
2-Iodo-5-methoxyaniline 97%
2-Iodo-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈INO
MDL Number: MFCD08458361
CAS Number: 153898-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR