2-Chloro-1-methylpyridinium-p-toluenesulfonate 98%
Supplier: Ambeed
|
Warning
|
A441406-100G
A441406-25G
A441406-5G
77264-990EA
62
USD
77264-990
77264-992
77264-994
2-Chloro-1-methylpyridinium-p-toluenesulfonate 98%
2-Chloro-1-methylpyridinium-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₁₃H₁₄ClNO₃S |
MDL Number:
MFCD00011985 CAS Number: 7403-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...