6-Amino-4-methylnicotinonitrile 97%

Supplier: Ambeed
Warning

A441321-1G A441321-250MG A441321-5G
77252-618EA 36 USD
77252-618 77252-620 77551-328
6-Amino-4-methylnicotinonitrile 97%
6-Amino-4-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD09839255
CAS Number: 179555-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR