(R)-2,2'-Dihydroxy-[1,1'-binaphthalene]-3,3'-dicarboxaldehyde ≥98%, ee 99%

Supplier: Ambeed
Warning

A440940-100MG A440940-250MG
77252-608EA 51 USD
77252-608 77252-610
(R)-2,2'-Dihydroxy-[1,1'-binaphthalene]-3,3'-dicarboxaldehyde ≥98%, ee 99%
(R)-2,2'-Dihydroxy-[1,1'-binaphthalene]-3,3'-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₂₂H₁₄O₄
MDL Number: MFCD27951997
CAS Number: 121314-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR