4-Methyl-1,3-oxazole-5-carbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-4-methyl-1,3-oxazole, 4-Methyloxazole-5-carbonitrile

A440149-100MG A440149-1G A440149-250MG
77291-600EA 51 USD
77291-600 77291-602 77291-604
4-Methyl-1,3-oxazole-5-carbonitrile 95%
4-Methyl-1,3-oxazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₂O
MDL Number: MFCD01111976
CAS Number: 1003-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR