DSP (Dithiobis(succinimidyl propionate)) 97%

Supplier: Ambeed
Warning

Synonyms: DTSSP, 3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), DSP Crosslinker, Lomant’s reagent, 3,3'-Dithiodipropionic acid bis(N-succinimidyl Ester), Di(N-succinimidyl)-3,3'-dithiodipropionate, Sulfo-DSP, DTSP, 3,3'-Dithiodipropionic acid di(N-succinimidyl Ester)

A440058-100MG A440058-1G A440058-250MG A440058-5G A440058-25G
77279-112EA 6.18 USD
77279-112 77279-114 77279-116 77616-892 77748-898
DSP (Dithiobis(succinimidyl propionate)) 97%
DSP (Dithiobis(succinimidyl propionate))

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₈S₂
MW: 404.41 g/mol
MDL Number: MFCD00042045
CAS Number: 57757-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR