4,6-Dichloro-2-(methylthio)pyrimidine 98%

Supplier: Ambeed
Danger

A440012-10G A440012-25G
77291-596EA 5 USD
77291-596 77291-598
4,6-Dichloro-2-(methylthio)pyrimidine 98%
4,6-Dichloro-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂S
MDL Number: MFCD00006086
CAS Number: 6299-25-8
UN: 3263
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR