4-Methyl-1H-pyrazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Fomepizole HCl

A439984-1G A439984-250MG
77291-592EA 262.65 USD
77291-592 77291-594
4-Methyl-1H-pyrazole hydrochloride 95%
4-Methyl-1H-pyrazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₂
MW: 118.5648 g/mol
MDL Number: MFCD00012707
CAS Number: 56010-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR