(4-Isobutoxyphenyl)methanamine acetate 98%

Supplier: Ambeed
Warning

A439717-100G A439717-1G A439717-25G A439717-5G
77279-104EA 215.27 USD
77279-104 77279-106 77279-108 77279-110
(4-Isobutoxyphenyl)methanamine acetate 98%
(4-Isobutoxyphenyl)methanamine acetate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
MW: 239.31 g/mol
MDL Number: MFCD30718115
CAS Number: 955997-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR