4,4',4''-(Pyridine-2,4,6-triyl)trianiline 97%

Supplier: Ambeed
Warning

A439442-100MG A439442-1G A439442-250MG A439442-5G
77264-966EA 35 USD
77264-966 77264-968 77264-970 77264-972
4,4',4''-(Pyridine-2,4,6-triyl)trianiline 97%
4,4',4''-(Pyridine-2,4,6-triyl)trianiline

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₄
MDL Number: MFCD31413128
CAS Number: 83266-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR