2-(2,4-Dimethylphenyl)pyridine 97%

Supplier: Ambeed
Warning

A439347-100MG A439347-1G A439347-250MG
77240-956EA 40.17 USD
77240-956 77240-958 77240-960
2-(2,4-Dimethylphenyl)pyridine 97%
2-(2,4-Dimethylphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.24 g/mol
MDL Number: MFCD07775645
CAS Number: 914253-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR