(R)-(-)-α-Methoxyphenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-alpha-Methoxyphenylacetic Acid

A438900-100G A438900-10G A438900-1G A438900-25G A438900-500G A438900-5G
77252-592EA 269.86 USD
77252-592 77252-594 77252-596 77252-598 77252-600 77252-602
(R)-(-)-α-Methoxyphenylacetic acid 98%
(R)-(-)-α-Methoxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00004250
CAS Number: 3966-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR