2,4-Dibromophenol 98%

Supplier: Ambeed
Danger

A438685-100G A438685-25G A438685-5G
77252-586EA 48 USD
77252-586 77252-588 77252-590
2,4-Dibromophenol 98%
2,4-Dibromophenol

Precaución: for Research Use Only

Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Melting Pt: 38
MDL Number: MFCD00002149
CAS Number: 615-58-7
UN: 2811
ADR: 6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR