N-(2,3,5,6-Tetrafluorophenyl)acetamide 95+%

Supplier: Ambeed
Warning

A438399-100MG A438399-1G A438399-250MG
77252-580EA 26 USD
77252-580 77252-582 77252-584
N-(2,3,5,6-Tetrafluorophenyl)acetamide 95+%
N-(2,3,5,6-Tetrafluorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₅F₄NO
MDL Number: MFCD00039213
CAS Number: 1766-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR