5,5-Dimethyl-5,13-dihydrobenzofuro[3,2-c]acridine 97%

Supplier: Ambeed
Warning

A438396-100MG A438396-1G A438396-250MG
77252-574EA 25 USD
77252-574 77252-576 77252-578
5,5-Dimethyl-5,13-dihydrobenzofuro[3,2-c]acridine 97%
5,5-Dimethyl-5,13-dihydrobenzofuro[3,2-c]acridine

Precaución: for Research Use Only

Formula: C₂₁H₁₇NO
MDL Number: MFCD30543391
CAS Number: 1381986-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR