3,4-(Ethylenedioxy)benzylamine 98%

Supplier: Ambeed
Warning

A438157-1G A438157-250MG
77252-570EA 43 USD
77252-570 77252-572
3,4-(Ethylenedioxy)benzylamine 98%
3,4-(Ethylenedioxy)benzylamine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD02677684
CAS Number: 17413-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR