2'-Acetoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetylphenyl acetate, 2′-Acetoxyacetophenone, o-Acetylphenyl acetate

A438113-100G A438113-25G
77252-566EA 119 USD
77252-566 77252-568
2'-Acetoxyacetophenone 98%
2'-Acetoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD00017223
CAS Number: 7250-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR