1-Bromo-2-butanol 70%

Supplier: Ambeed
Danger

A437258-1G A437258-250MG A437258-5G
77240-936EA 22 USD
77240-936 77240-938 77746-892
1-Bromo-2-butanol 70%
1-Bromo-2-butanol

Precaución: for Research Use Only

Formula: C₄H₉BrO
MDL Number: MFCD00059883
CAS Number: 2482-57-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR