3-Methoxyisonicotinonitrile 97%

Supplier: Ambeed
Warning

A437241-250MG A437241-5G A437241-1G
77240-934EA 19.57 USD
77240-934 77616-880 77746-891
3-Methoxyisonicotinonitrile 97%
3-Methoxyisonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.13 g/mol
MDL Number: MFCD13175841
CAS Number: 26414-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR