4-Bromo-2,3-dimethylphenol 98%

Supplier: Ambeed
Warning

A437084-1G A437084-250MG
77291-584EA 49.44 USD
77291-584 77291-586
4-Bromo-2,3-dimethylphenol 98%
4-Bromo-2,3-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD07778969
CAS Number: 22802-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR