5,7-Dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-4H-chromen-4-one 95%

Supplier: Ambeed
Warning

Synonyms: Tricin

A436856-10MG A436856-1MG A436856-2MG A436856-50MG A436856-5MG
77279-084EA 66 USD
77279-084 77279-086 77279-088 77748-887 77748-888
5,7-Dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-4H-chromen-4-one 95%
5,7-Dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₇
MDL Number: MFCD00210580
CAS Number: 520-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR