2,4-Dibromoanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dibromo-4-methoxybenzene, 2,4-Dibromo-1-methoxybenzene

A436840-100G A436840-25G A436840-500G A436840-5G
77252-550EA 50.47 USD
77252-550 77252-552 77252-554 77252-556
2,4-Dibromoanisole 98%
2,4-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
MDL Number: MFCD00000079
CAS Number: 21702-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR