2-Amino-1-methylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-benzo[d]imidazol-2-amine

A436806-250MG A436806-5G A436806-1G
77252-546EA 17.51 USD
77252-546 77252-548 77551-310
2-Amino-1-methylbenzimidazole 98%
2-Amino-1-methylbenzimidazole

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.181 g/mol
MDL Number: MFCD00142855
CAS Number: 1622-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR